Assay Details
| Gene Symbol: ebp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075125.6 | NP_507526.1 | 2 - 3 | 330 | 129 | |
| Gene Symbol: ebp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075127.3 | NP_507528.1 | 2 - 3 | 332 | 139 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_075127.3 transcripts.
Set Membership: |
|

ebp-1
Genomic Map