Assay Details
| Gene Symbol: oac-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075335.4 | NP_507736.3 | 8 - 9 | 1381 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

oac-49| Gene Symbol: oac-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075335.4 | NP_507736.3 | 8 - 9 | 1381 | 83 | |
Set Membership: |
|