Assay Details
| Gene Symbol: lgc-55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075469.6 | NP_507870.2 | 5 - 6 | 759 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lgc-55| Gene Symbol: lgc-55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075469.6 | NP_507870.2 | 5 - 6 | 759 | 86 | |
Set Membership: |
|