Assay Details
| Gene Symbol: tmc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075820.8 | NP_508221.3 | 20 - 21 | 3047 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tmc-1| Gene Symbol: tmc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075820.8 | NP_508221.3 | 20 - 21 | 3047 | 90 | |
Set Membership: |
|