Assay Details
| Gene Symbol: kin-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076092.6 | NP_508493.1 | 10 - 11 | 1313 | 80 | |
| GenBank mRNA | AF403714.1 | - | 10 - 11 | 1245 | 80 | |
More Information
Set Membership: |
|

kin-29
Genomic Map