Assay Details
| Gene Symbol: kin-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076092.6 | NP_508493.1 | 15 - 16 | 2416 | 128 | |
| GenBank mRNA | AF403714.1 | - | 15 - 16 | 2348 | 128 | |
More Information
Set Membership: |
|

kin-29
Genomic Map