Assay Details
| Gene Symbol: kin-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076092.6 | NP_508493.1 | 7 - 8 | 850 | 76 | |
| GenBank mRNA | AF403714.1 | - | 7 - 8 | 782 | 76 | |
More Information
Set Membership: |
|

kin-29
Genomic Map