Assay Details
| Gene Symbol: apl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076469.6 | NP_508870.3 | 11 - 12 | 1739 | 65 | |
| NM_076470.7 | NP_508871.1 | 11 - 12 | 1735 | 65 | ||
More Information
Set Membership: |
|

apl-1
Genomic Map