Assay Details
| Gene Symbol: apl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076469.6 | NP_508870.3 | 1 - 2 | 67 | 66 | |
| NM_076470.7 | NP_508871.1 | 1 - 2 | 69 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

apl-1| Gene Symbol: apl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076469.6 | NP_508870.3 | 1 - 2 | 67 | 66 | |
| NM_076470.7 | NP_508871.1 | 1 - 2 | 69 | 66 | ||
Set Membership: |
|