Assay Details
| Gene Symbol: apl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076469.6 | NP_508870.3 | 6 - 7 | 819 | 91 | |
| NM_076470.7 | NP_508871.1 | 6 - 7 | 821 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

apl-1| Gene Symbol: apl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076469.6 | NP_508870.3 | 6 - 7 | 819 | 91 | |
| NM_076470.7 | NP_508871.1 | 6 - 7 | 821 | 91 | ||
Set Membership: |
|