Assay Details
| Gene Symbol: dpyd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076526.6 | NP_508927.2 | 8 - 9 | 1384 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpyd-1| Gene Symbol: dpyd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076526.6 | NP_508927.2 | 8 - 9 | 1384 | 71 | |
Set Membership: |
|