Assay Details
| Gene Symbol: twnk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368518.5 | NP_001355545.1 | 12 | 1466 | 83 | |
| NM_001380982.1 | NP_001368724.1 | 11 - 12 | 1494 | 83 | ||
| NM_001383588.1 | 13 - 14 | 1792 | 83 | |||
| NM_076572.6 | NP_508973.3 | 12 - 13 | 1508 | 83 | ||
More Information
Set Membership: |
|

twnk-1
Genomic Map