Assay Details
| Gene Symbol: twnk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368518.5 | NP_001355545.1 | 7 | 888 | 59 | |
| NM_001380982.1 | NP_001368724.1 | 5 - 6 | 862 | 59 | ||
| NM_001383588.1 | 8 - 9 | 1214 | 59 | |||
| NM_076572.6 | NP_508973.3 | 6 - 7 | 849 | 59 | ||
More Information
Set Membership: |
|

twnk-1
Genomic Map