Assay Details
| Gene Symbol: ipp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361812.4 | NP_001348790.1 | 1 - 2 | 95 | 65 | |
| NM_076607.8 | NP_509008.2 | 1 - 2 | 95 | 65 | ||
More Information
Set Membership: |
|

ipp-5
Genomic Map