Assay Details
| Gene Symbol: ipp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361812.4 | NP_001348790.1 | 2 - 3 | 278 | 91 | |
| NM_076607.8 | NP_509008.2 | 2 - 3 | 278 | 91 | ||
More Information
Set Membership: |
|

ipp-5
Genomic Map