Assay Details
| Gene Symbol: ist-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330903.3 | NP_001317892.1 | 14 - 15 | 2363 | 113 | |
| NM_001373345.2 | NP_001360631.1 | 16 - 17 | 2645 | 113 | ||
| NM_001373346.2 | NP_001360716.1 | 15 | 2375 | 113 | ||
| NM_076800.7 | NP_509201.3 | 13 - 14 | 2145 | 113 | ||
More Information
Set Membership: |
|

ist-1
Genomic Map