Assay Details
| Gene Symbol: ist-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330903.3 | NP_001317892.1 | 11 - 12 | 1745 | 125 | |
| NM_001373345.2 | NP_001360631.1 | 13 - 14 | 2027 | 125 | ||
| NM_001373346.2 | NP_001360716.1 | 12 | 1757 | 125 | ||
| NM_076800.7 | NP_509201.3 | 10 - 11 | 1527 | 125 | ||
More Information
Set Membership: |
|

ist-1
Genomic Map