Assay Details
| Gene Symbol: lgc-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076869.6 | NP_509270.1 | 3 - 4 | 507 | 94 | |
| GenBank mRNA | AY519852.1 | - | 3 - 4 | 507 | 94 | |
More Information
Set Membership: |
|

lgc-11
Genomic Map