Assay Details
| Gene Symbol: adt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029361.5 | NP_001024532.2 | 3 - 4 | 190 | 152 | |
| NM_001375070.2 | NP_001361970.1 | 2 | 190 | 152 | ||
| GenBank mRNA | AB072347.1 | - | 3 - 4 | 190 | 152 | |
More Information
Set Membership: |
|

adt-2
Genomic Map