Assay Details
| Gene Symbol: tbc-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077020.7 | NP_509421.2 | 11 - 12 | 1681 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tbc-18| Gene Symbol: tbc-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077020.7 | NP_509421.2 | 11 - 12 | 1681 | 96 | |
Set Membership: |
|