Assay Details
| Gene Symbol: tbc-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077020.7 | NP_509421.2 | 14 - 15 | 2383 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tbc-18| Gene Symbol: tbc-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077020.7 | NP_509421.2 | 14 - 15 | 2383 | 109 | |
Set Membership: |
|