Assay Details
| Gene Symbol: lgc-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077155.5 | NP_509556.4 | 8 - 9 | 1676 | 149 | |
| Gene Symbol: col-175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383621.1 | 6 - 7 | 1042 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383621.1 transcripts.
Set Membership: |
|

lgc-4
Genomic Map