Assay Details
| Gene Symbol: oga-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129711.3 | NP_001123183.1 | 7 - 8 | 1090 | 67 | |
| NM_001129712.4 | NP_001123184.1 | 6 - 7 | 1140 | 67 | ||
| GenBank mRNA | DQ407522.1 | - | 7 - 8 | 1040 | 67 | |
More Information
Set Membership: |
|

oga-1
Genomic Map