Assay Details
| Gene Symbol: flp-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077375.6 | NP_509776.1 | 2 - 3 | 312 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

flp-19| Gene Symbol: flp-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077375.6 | NP_509776.1 | 2 - 3 | 312 | 71 | |
Set Membership: |
|