Assay Details
| Gene Symbol: abl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381086.2 | NP_001368776.1 | 10 - 11 | 2408 | 135 | |
| NM_001381087.2 | NP_001368775.1 | 12 - 13 | 2498 | 135 | ||
| NM_077376.10 | NP_509777.2 | 11 - 12 | 2469 | 135 | ||
| NM_077377.6 | NP_509778.2 | 12 - 13 | 2489 | 135 | ||
| NM_077378.5 | NP_509779.2 | 8 - 9 | 2546 | 135 | ||
More Information
Set Membership: |
|

abl-1
Genomic Map