Assay Details
| Gene Symbol: abl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381086.2 | NP_001368776.1 | 1 - 2 | 56 | 85 | |
| NM_001381087.2 | NP_001368775.1 | 2 - 3 | 116 | 85 | ||
| NM_077376.10 | NP_509777.2 | 2 - 3 | 117 | 85 | ||
| NM_077377.6 | NP_509778.2 | 2 - 3 | 107 | 85 | ||
More Information
Set Membership: |
|

abl-1
Genomic Map