Assay Details
| Gene Symbol: abl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381086.2 | NP_001368776.1 | 2 - 3 | 160 | 64 | |
| NM_077376.10 | NP_509777.2 | 3 - 4 | 221 | 64 | ||
More Information
Set Membership: |
|

abl-1
Genomic Map