Assay Details
| Gene Symbol: abl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381086.2 | NP_001368776.1 | 3 - 4 | 232 | 73 | |
| NM_001381087.2 | NP_001368775.1 | 5 - 6 | 322 | 73 | ||
| NM_077376.10 | NP_509777.2 | 4 - 5 | 293 | 73 | ||
| NM_077377.6 | NP_509778.2 | 5 - 6 | 313 | 73 | ||
More Information
Set Membership: |
|

abl-1
Genomic Map