Assay Details
| Gene Symbol: abl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381086.2 | NP_001368776.1 | 5 - 6 | 725 | 91 | |
| NM_001381087.2 | NP_001368775.1 | 7 - 8 | 815 | 91 | ||
| NM_077376.10 | NP_509777.2 | 6 - 7 | 786 | 91 | ||
| NM_077377.6 | NP_509778.2 | 7 - 8 | 806 | 91 | ||
| NM_077378.5 | NP_509779.2 | 3 - 4 | 863 | 91 | ||
More Information
Set Membership: |
|

abl-1
Genomic Map