Assay Details
| Gene Symbol: abl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381087.2 | NP_001368775.1 | 4 - 5 | 241 | 66 | |
| NM_077377.6 | NP_509778.2 | 4 - 5 | 232 | 66 | ||
More Information
Set Membership: |
|

abl-1
Genomic Map