Assay Details
| Gene Symbol: ver-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077434.4 | NP_509835.2 | 11 - 12 | 3264 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ver-4| Gene Symbol: ver-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077434.4 | NP_509835.2 | 11 - 12 | 3264 | 136 | |
Set Membership: |
|