Assay Details
| Gene Symbol: ldb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381099.1 | NP_001366942.1 | 2 - 3 | 238 | 94 | |
| NM_077444.6 | NP_509845.1 | 2 - 3 | 195 | 94 | ||
More Information
Set Membership: |
|

ldb-1
Genomic Map