Assay Details
| Gene Symbol: ldb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381099.1 | NP_001366942.1 | 4 - 5 | 465 | 112 | |
| NM_001381100.2 | NP_001366860.1 | 5 - 6 | 695 | 112 | ||
| NM_077444.6 | NP_509845.1 | 4 - 5 | 422 | 112 | ||
| GenBank mRNA | U80220.1 | - | 3 - 4 | 354 | 112 | |
More Information
Set Membership: |
|

ldb-1
Genomic Map