Assay Details
| Gene Symbol: ldb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381099.1 | NP_001366942.1 | 6 - 7 | 923 | 92 | |
| NM_001381100.2 | NP_001366860.1 | 7 - 8 | 1153 | 92 | ||
| NM_077444.6 | NP_509845.1 | 6 - 7 | 880 | 92 | ||
| GenBank mRNA | U80220.1 | - | 5 - 6 | 812 | 92 | |
More Information
Set Membership: |
|

ldb-1
Genomic Map