Assay Details
| Gene Symbol: ldb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381099.1 | NP_001366942.1 | 7 - 8 | 1001 | 65 | |
| NM_001381100.2 | NP_001366860.1 | 8 - 9 | 1231 | 65 | ||
| NM_077444.6 | NP_509845.1 | 7 - 8 | 958 | 65 | ||
| GenBank mRNA | U80220.1 | - | 6 - 7 | 890 | 65 | |
More Information
Set Membership: |
|

ldb-1
Genomic Map