Assay Details
| Gene Symbol: ldb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381100.2 | NP_001366860.1 | 3 - 4 | 474 | 83 | |
| GenBank mRNA | U80220.1 | - | 1 - 2 | 133 | 83 | |
More Information
Set Membership: |
|

ldb-1
Genomic Map