Assay Details
| Gene Symbol: gcy-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077496.5 | NP_509897.3 | 12 - 13 | 1864 | 79 | |
| GenBank mRNA | HQ636455.1 | - | 11 - 12 | 1473 | 79 | |
More Information
Set Membership: |
|

gcy-9
Genomic Map