Assay Details
| Gene Symbol: gcy-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077496.5 | NP_509897.3 | 19 - 20 | 2874 | 114 | |
| GenBank mRNA | HQ636455.1 | - | 18 - 19 | 2483 | 114 | |
More Information
Set Membership: |
|

gcy-9
Genomic Map