Assay Details
| Gene Symbol: selt-1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077528.4 | NP_509929.2 | 4 - 5 | 401 | 64 | |
| NM_077529.5 | NP_509930.2 | 4 - 5 | 392 | 64 | ||
More Information
Set Membership: |
|

selt-1.1
Genomic Map