Assay Details
| Gene Symbol: pri-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268086.2 | NP_001255015.1 | 6 - 7 | 1084 | 106 | |
| NM_001268087.3 | NP_001255016.1 | 2 - 3 | 203 | 106 | ||
More Information
Set Membership: |
|

pri-1
Genomic Map