Assay Details
| Gene Symbol: TENT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618175.3 | XP_005618232.1 | 774 | 89 | ||
| XM_005618176.3 | XP_005618233.1 | 773 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TENT2
Genomic Map