Assay Details
| Gene Symbol: FOXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639326.4 | XP_003639374.1 | 2 - 3 | 697 | 110 | |
| XM_014117518.2 | XP_013972993.1 | 2 - 3 | 659 | 110 | ||
| XM_022424692.1 | XP_022280400.1 | 2 - 3 | 727 | 110 | ||
More Information
Set Membership: |
|

FOXN2
Genomic Map