Assay Details
| Gene Symbol: FBXO28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421208.1 | XP_022276916.1 | 1 - 2 | 295 | 71 | |
| XM_849736.5 | XP_854829.1 | 1 - 2 | 295 | 71 | ||
More Information
Set Membership: |
|

FBXO28
Genomic Map