Assay Details
| Gene Symbol: UBE2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408334.1 | XP_022264042.1 | 422 | 71 | ||
| XM_850890.5 | XP_855983.1 | 600 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UBE2D2
Genomic Map