Assay Details
| Gene Symbol: UBE2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628390.3 | XP_005628447.1 | 4 - 5 | 772 | 64 | |
| XM_022427231.1 | XP_022282939.1 | 6 - 7 | 513 | 64 | ||
| XM_022427234.1 | XP_022282942.1 | 6 - 7 | 589 | 64 | ||
| XM_022427235.1 | XP_022282943.1 | 5 - 6 | 557 | 64 | ||
| XM_852326.5 | XP_857419.1 | 6 - 7 | 862 | 64 | ||
More Information
Set Membership: |
|

UBE2H
Genomic Map