Assay Details
| Gene Symbol: TLR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002950.2 | NP_001002950.2 | 3 | 3014 | 120 | |
| GenBank mRNA | AB080363.1 | - | 1 | 2609 | 120 | |
More Information
Set Membership: |
|

TLR4
Genomic Map