Assay Details
| Gene Symbol: FCGR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002976.2 | NP_001002976.1 | 1 - 2 | 88 | 85 | |
| XM_022404314.1 | XP_022260022.1 | 2 - 3 | 527 | 88 | ||
| GenBank mRNA | AB101519.1 | - | 1 - 2 | 88 | 85 | |
More Information
Set Membership: |
|

FCGR1A
Genomic Map