Assay Details
| Gene Symbol: CCL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005250.1 | NP_001005250.1 | 1 - 2 | 78 | 74 | |
| GenBank mRNA | AB163435.1 | - | 1 - 2 | 78 | 74 | |
| AB183194.1 | - | 1 - 2 | 78 | 74 | ||
More Information
Set Membership: |
|

CCL4
Genomic Map