Assay Details
| Gene Symbol: CCL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010960.1 | NP_001010960.1 | 2 - 3 | 190 | 98 | |
| GenBank mRNA | AB183286.1 | - | 2 - 3 | 190 | 98 | |
| AY849795.1 | - | 2 - 3 | 190 | 98 | ||
More Information
Set Membership: |
|

CCL7
Genomic Map