Assay Details
| Gene Symbol: GUSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003191.1 | NP_001003191.1 | 1849 | 75 | ||
| XM_005620897.3 | XP_005620954.1 | 1801 | 75 | |||
| XM_014114125.2 | XP_013969600.1 | 1352 | 75 | |||
| XM_014114126.2 | XP_013969601.1 | 1133 | 75 | |||
| GenBank mRNA | AF019759.1 | - | 1849 | 75 | ||
More Information
Set Membership: |
|

GUSB
Genomic Map